Deckgläser, rund
Lieferant: MARIENFELD
0111540 0111530 0111700 0111580 0111560 0111600 0111620 0111640 0111500 0111520 0111550 0111650
631-0692EA 66.9 EUR
631-0692 630-1597 631-0694 631-0669 631-0668 631-0670 631-0671 631-0672 631-0665 631-0666 631-0667 MARI0111650
Deckgläser, rund
Deckgläser
Round cover glasses made of pure white borosilicate D 263 ™ M glass, Hydrolytic Class I.
  • No bubbles or flaws
Suitable for in vitro diagnostic (IVD) applications.
Verpackung: 100 cover glasses per box with separate lid.