Slide holder, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F445420000
631-9108EA 46.8 EUR
631-9108
Slide holder, SP Bel-Art
Objektträgeraufbewahrung
Slide holder designed to accomodate up to ten slides.
  • Plastic durable construction for regular use
  • Inclined channel to hold slides vertically
  • Front ledge for drying slides