Färbeküvette
Lieferant: KARTELL
351
631-9131EA 127 EUR
631-9131
Färbeküvette
Dosen Färbeküvetten
Made from PMP (TPX).
This rectangular jar is supplied with a lid and can hold 10 slides or 20 slides back-to-back.