Economical Storage Box
Lieferant: DELTALAB
DS42
631-9790EA 457 EUR
631-9790
Economical Storage Box
Boxen Lagerboxen
Made of heavy-duty cardboard for long term storage.
This box, complete with a lid, is specially designed for the storage of histology cassettes.
Each box stores up to 250 cassettes or 160 embedding rings.