Staining jar and basket
Lieferant: DELTALAB
19360
631-9810EA 208 EUR
631-9810
Staining jar and basket
Dosen Färbeküvetten
Made of stainless steel, with lid and adjustable handle.
  • With lid to avoid evaporation
  • Excellent chemical resistance
  • Excellent temperature resistance
The staining basket hold up to 30 slides (76×26 mm).