Magnesium ionophore VI, Selectophore™, Funktion getestet Ionophor, Supelco®
Lieferant: Merck
Synonyme:
1,3,5-Tris[10-(1-adamantyl)-7,9-dioxo-6,10-diazaundecyl]benzene, ETH 5506
M-Clarity™ Qualitätsstufe = MQ100
63112-10MG
63112-100MG
63112-10MGEA
1180
EUR
63112-10MG
63112-100MG
Magnesium ionophore VI, Selectophore™, Funktion getestet Ionophor, Supelco®
Magnesium ionophore VI
Lipophilic magnesium-selective ionophore (log PTLC=8.5) with sufficient selectivity for Mg2+ against the main interfering cations for serum measurements.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₆₃H₉₆N₆O₆ Molecular Weight: 1033.47 g/mol |
MDL:
MFCD01867417 CAS-Nummer: 151058-38-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...