Magnesium ionophore VI, Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck

Synonyme: 1,3,5-Tris[10-(1-adamantyl)-7,9-dioxo-6,10-diazaundecyl]benzene, ETH 5506

M-Clarity™ Qualitätsstufe = MQ100

63112-10MG 63112-100MG
63112-10MGEA 1180 EUR
63112-10MG 63112-100MG
Magnesium ionophore VI, Selectophore™, Funktion getestet Ionophor, Supelco®
Magnesium ionophore VI

Lipophilic magnesium-selective ionophore (log PTLC=8.5) with sufficient selectivity for Mg2+ against the main interfering cations for serum measurements.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₆₃H₉₆N₆O₆
Molecular Weight: 1033.47 g/mol
MDL: MFCD01867417
CAS-Nummer: 151058-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09096dd4708b56e47d923dfc45548236331d232e&vid=25dffe7713b51325b544cc5964b6756db442384c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR