Microtitre Plates, Black, 96-Well, Sterilin®
Lieferant: Sterilin
611F96BK
634-0006EA 233 EUR
634-0006
Microtitre Plates, Black, 96-Well, Sterilin®
Mikrotiterplatten HTS-Mikrotiterplatten
PS, non-treated. Ideal for fluorescence work and bioluminescence studies.
  • Black 96-well plates are ideal for fluorescence work where the pigment prevents cross-talk between the wells - low background fluorescence minimises light scattering
  • SBS compliant, ensuring suitable fit with all automation
  • Manufactured from virgin polystyrene
  • Supplied nonsterile, providing a medium binding surface
  • Chimney well design to help reduce contamination
  • Supplied with no lid
Farbe: Schwarz