KÜVETTE 115-QS 10MM 400µL SPEKTRAL GEM. 1 * 1 PAAR
Lieferant: HELLMA
Micro cuvette, with PTFE stopper, Micro, QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm, Path length: 10 mm, Height: 40 mm, Volume: 400 µl