UV/Vis Spectrophotometers, Ultrospec™ 9000
Lieferant: BIOCHROM
29003619
634-0891EA 0 EUR
634-0891
UV/Vis Spectrophotometers, Ultrospec™ 9000
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
UV/Vis spectrophotometers designed to meet the requirements of critical analysis and work in controlled or regulated laboratory environments. Both stand-alone and computer-controlled models are available for both series.
  • Fully complies with the requirements of the European pharmacopoeia
  • Clear and easy to use color touch screen (stand alone device models)
  • Apply custom calculations to your measurement results
  • Multi-user analytics device: Secure login to ensure secure methods
  • The delivery includes software that includes the following functions: wavelength spectral measurement, fixed wavelength, standard curves, kinetics, and pre-recorded bioscientific methods