VWR®, Makroküvetten
Lieferant: VWR Collection
634-1067EA 21.6 EUR
634-1067
VWR®, Makroküvetten
Küvetten Spektralphotometer-Küvetten
Einweg-Makroküvetten, mit vier durchsichtigen Seiten.
  • Clear and clean optical path
  • Four windows for precise measurements
  • Manufactured under controlled room conditions
Specially designed for fluorescence spectroscopy.