UV-Visible/NIR Spectrophotometer, UH5700
Lieferant: VWR Collection
2J3-0013
634-1242EA 26770 EUR
634-1242
UV-Visible/NIR Spectrophotometer, UH5700
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
The UH5700 UV-Vis-NIR Spectrophotometer is a high-precision benchtop spectrophotometer instrument designed for handling the ultraviolet, visible and near-infrared regions (190 to 3300 nm).
  • Continuously variable slit
  • Low stray light/high photometric range
  • Supports high-speed scanning
  • New software UV solutions plus
  • Multiple accessories for a wide range of measurements
UH5700 achieves best-in-class levels of low stray light and high photometric range through use of a Czerny-Turner mounted single-monochromator bright spectrometer and a newly developed grating. The UH5700 employs a continuously variable slit in the near-infrared region, in which the slit automatically widens when measuring low quantities of light and narrows when measuring large quantities of light, and thereby achieves low-noise measurements across a broad range of measurement wavelengths.
The sample compartment shares a common design with the U-2900/U-3900, so existing accessories can be used (Except for certain accessories). Please check with your Avantor sales representative for details).
Software:
UH5700 UV-Visible/NIR Spectrophotometer utilises the UV Solutions Plus software for instrument control and data processing. Enhancements have been added compared to the popular UV Solutions Software (predecessor model) including a tabular form display feature for data lists and data processing results, a report layout feature, a performance confirmation feature.
Zubehörinformationen:
For measurement of liquids
Micro cell option
Micro cells are used in combination with a mask (HITA200-1537). A micro cell is suitable for small samples with volumes of 340 to 600 µl.
Ultra-micro volume sample measurement option
Trace samples are used in combination with a mask (HITA3J1-0116) (two included). These are suitable for ultra-micro volumes 1,5 to 90 µl.
Autosipper (HITA2J3-0131)
Suitable for multiple sample measurements.
AS-1010 Autosampler (HITA2J1-0122)
In combination with auto sipper, autosampler can make multiple automatic measurement of solution samples.
For measurement of solids
Ø60 Integrating Sphere (HITA2J3-0176)
Used for absorbance measurements of turbid samples and reflectance measurement of solid sample surfaces.
5° Specular Reflectance Accessory (relative)(HITA2J3-0151)
This accessory uses the mirror reflection of a sample to measure relative reflectance with respect to a standard reflection plate (aluminum-evaporated plane mirror). This is used for film thickness measurement, spectral reflectance measurement, and other such applications.
Glass filter holder (HITA210-2109)
Used for measuring transmittance and absorbance of solid plate samples such as glass filters.
Film holder (HITA210-2112)
Used for measuring the transmittance and absorbance of film-like samples.
Bestellinformation:
Please contact your local Avantor Sales representative for more information on range of accessories.
Lieferumfang:
The UH5700 spectrophotometer is delivered with USB communication cable, instruction manual, UV solutions plus software program.