UV/VIS Spectrophotometers
Lieferant: VWR Collection
2J1-0033 2J1-0023
634-1244EA 9260 EUR
634-1244 634-1243
UV/VIS Spectrophotometers
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
The UH5200 features a 10,4” backlit colour LCD display with 800×600-pixel resolution to allow detailed visualisation of calibration curves and measured spectra. The UH5200 is a standalone instrument that may also be controlled from a PC by installing the optional UV Solutions Plus software package. Measured data may be stored in the instrument´s on-board memory or on USB drives.
  • Large colour LCD and safe input with keypad
  • Supports barcode reader for standalone use
  • Data storage via USB supported
  • New lamp controls designed for safety and environmental impact
  • Automated calibration and self-diagnostic capabilities
UH5200 series instruments are equipped with the capability to turn the lamp on and off automatically, thus avoiding unnecessary on-time. When the lamp is turned on, the instrument monitors its output and automatically notifies the operator when this output has stabilised, indicating that measurements may now begin. Also, as a new safety provision, the UH5200 series now incorporates a sensor to detect whether the lamp chamber is open or closed during the lamp-replacement process.
The spectrophotometer can connect to a commercially available printer through its USB printer port.
The UH5200 incorporates a variety of automated calibration and self-diagnostic capabilities-including memory checks, wavelength-drive checks, illumination checks, wavelength auto-calibration, and a lamp on-time display feature-to ensure stable operation of the instrument.
The UH5210 is a fully PC-controlled instrument. Its control software is UV Solutions Plus, a new platform that is an update to the UV Solutions control software of the previous-generation U-2900 system with expanded functionality, including display of data lists and data-processing results in tabular form, report layouts, and performance validation. 
The UH5210 is a successor to the U-2910 and is a PC-controlled model inheriting its robustness and comfortable operability.