VWR®, V-3000PC, Vis-Spektralphotometer
Lieferant: VWR Collection
634-6040EA 3010 EUR
634-6040
VWR®, V-3000PC, Vis-Spektralphotometer
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
The V-3000 is a highly robust unit designed for advanced routine measurements and educational applications.
  • Easy to use
  • USB port for result export
  • Photometrics (absorption, transmission)
  • Quantitative, kinetics
  • Wavelength scan, multi-wavelength measurement
Including PC software.
Lieferumfang: Supplied with manual 4-position cell changer, 4 optical glass cuvettes (10×10mm), software and USB cable for PC control, USB stick, power cable with UK, CH and EU plug and dust cover.