Küvette 6030-OG, Optisches Glas, Schichtdicke 40 mm HELLMA 0 1 * 1 ST
Lieferant: HELLMA
Absorption cuvette, Macro, OG: Optical glass, for measurements in the Vis range from 360 - 2500 nm, Path length: 40 mm, Height: 45 mm, Volume: 14000 µl