Küvette 104.002B-QS, Quarzglas SUPRASIL, Schichtdicke 10 mm HELLMA 0 1 * 1 ST
Lieferant: HELLMA
Micro cuvette, with PTFE lid, Micro, QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm, Path length: 10 mm, Height: 45 mm, Volume: 700 µl