KÜVETTE 108.002-QS 10MM 500UL 1 * 1 ST
Lieferant: HELLMA
Micro cuvette, with PTFE lid, Micro, QS: Quartz glass, high performance for measurements in the UV/Vis range from 200 - 2500 nm, Path length: 10 mm, Height: 45 mm, Volume: 500 µl