Handheld refractometers
Lieferant: EUROMEX
RF.5580
635-0081EA 0 EUR
635-0081
Handheld refractometers
Refraktometer Handrefraktometer
Single dual or triple scales, analogue, handheld refractometers.
  • Automated temperature compensation (ATC)
  • Calibrated at 20 °C except for RF.5190
  • Requires only a few drops of liquid