Tragbare Refraktometer, digital, Kraftfahrzeugpflege, Chek-Serie
Lieferant: REICHERT
13940017
635-0510EA 599 EUR
635-0510
Tragbare Refraktometer, digital, Kraftfahrzeugpflege, Chek-Serie
Refraktometer Handrefraktometer
To measure the key fluids for automotive to help maintain safe operation on the road. Test brake fluids by measuring the boiling point to indicate the extent of the breakdown of the fluid or increase in water content.
  • Distilled water calibration
  • Glass prism
Standards: IP 65, RoHS and WEEE
Lieferumfang: Supplied with two AAA batteries for 10 000 readings.