Handheld refractometer, OPTi Digital, with 50 accessible scales
Lieferant: BELLINGHAM STANLEY
38-01
635-0832EA 391 EUR
635-0832
Handheld refractometer, OPTi Digital, with 50 accessible scales
Refraktometer Handrefraktometer
OPTi digital handheld refractometer with onboard library of 50 common scales including Brix and RI.
  • One instrument – 50 scales
  • Wide measuring range
  • Special features menu
  • Ergonomic IP 65 design
  • Truly portable for the laboratory or field
Packed full of scales and features, the pocket friendly OPTi refractometer has a wide measuring range covering the full 0 to 95 Brix range (1,33 to 1,53 RI). Up to three scales (A, B, C) can be easily activated from its onboard library of 50 scales at any time. Where applicable, Automatic Temperature Compensation (ATC) is applied to the displayed reading according to the scale or sample being measured.
Other features available through the OPTi refractometers special features menu includes -HAL- high ambient light detection, unique AG Test Mode for simple verification of performance, Delay before reading that helps stabilise the sample’s temperature prior to taking a reading and selection of temperature display parameter; all protected by a resettable PIN.
The OPTi refractometer provides all your measurement needs from a single instrument, whether that be measuring fruit, juice, confectionery, animal health, chemical composition and such like.
Standards: IP protection class according to DIN EN 60529: IP 65
Lieferumfang: Supplied with a soft carrying case and instructions.