Mineralöl schwer, weiß stabilisiert USP, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr


Stabilisierung: Stabilisiert mit Vitamin E 10-20 ppm

6357-10EA 246.78 EUR
6357-10 6357-19 6357-04
Mineralöl schwer, weiß stabilisiert USP, Macron Fine Chemicals™
Mineralöl
Dichte: 0,88 g/cm³
MDL: MFCD00131611
CAS-Nummer: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07263

Bestellen

Spezifikation Testergebnisse

Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
USP - Acidity Passes Test
USP - Limit of Polycyclic Aromatic Hydrocarbons Passes Test
USP - Limit of Sulfur Compounds Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Gravity at 25°/25°C 0.845 - 0.905
USP - Viscosity at 40 C, mm₂/s 34.5 - 150.0
USP - Readily Carbonizable Substances Passes Test
USP - Solid Paraffin Passes Test
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR