Organic reference standard, Ethyl acetoacetate, analytical standard, Supelco®
Lieferant: Merck
63954-1ML
63954-1MLEA 53.2 EUR
63954-1ML
Organic reference standard, Ethyl acetoacetate, analytical standard, Supelco®
Standards Organische Standards
Ethyl acetoacetate is derived from ethanol.
It can be used as a diluent for improving cold flow properties of biodiesel from waste cooking oil and as an additive for activating Fe₃O₄ nanoparticles for efficient catalytic oxidation of aldehydes to carboxylic acids.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=672d4743743d5906a20752368e5e27dc5760f0dc&vid=672d4743743d5906a20752368e5e27dc5760f0dc&prid=web&clid=SAPDAM