Organic reference standard, Ethyl acetoacetate, analytical standard, Supelco®
Lieferant: Merck
63954-1ML
63954-1MLEA
53.2
EUR
63954-1ML
Organic reference standard, Ethyl acetoacetate, analytical standard, Supelco®
Standards
Organische Standards
Ethyl acetoacetate is derived from ethanol.
It can be used as a diluent for improving cold flow properties of biodiesel from waste cooking oil and as an additive for activating Fe₃O₄ nanoparticles for efficient catalytic oxidation of aldehydes to carboxylic acids.