Sole-Tester
Lieferant: Alla France
1120FG026/15QP
641-1500EA 26.3 EUR
641-1500
Sole-Tester
Aräometer
Brine is the percentage of saturated solution of salt, usually sodium chloride, in water.
A brinometer is used to measure the percentage of saturated solution of brine.