Supelco® TraceCERT® Organic Reference Standard, Bromobutide
Lieferant: Merck
64148-50MG
64148-50MGEA 164 EUR
64148-50MG
Supelco® TraceCERT® Organic Reference Standard, Bromobutide
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9002f61464a3f96e3cc510bd6e71e453324ac5fd&vid=9002f61464a3f96e3cc510bd6e71e453324ac5fd&prid=web&clid=SAPDAM