Aceton ≥99,9%, CHROMASOLV™ Plus für die HPLC, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

650501-1L
650501-1LEA 74.79 EUR
650501-1L
Aceton ≥99,9%, CHROMASOLV™ Plus für die HPLC, Riedel-de Haën™
Aceton

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Formel: CH₃COCH₃
Molecular Weight: 58.08 g/mol
Siedepunkt: 55…57 °C (1.013 hPa)
Schmelzpunkt: –95 °C
Dichte: 0.79 g/cm³ (20 °C)
Flammpunkt: 14.3 %(V)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Refractive Index n20/D 1.359 (lit.)
Purity (GC) Min. 99.90 %
GC (as heptachlor epoxide) Max. 10 ng/l
Water (Karl Fischer) Max. 0.5 %
Non-volatile matter Max. 0.0003 %
IR spectrum complying
Absorbance at 400 nm Max. 0.005
Absorbance at 375 nm Max. 0.005
Absorbance at 350 nm Max. 0.01
Absorbance at 340 nm Max. 0.06
Absorbance at 330 nm Max. 1.0
Fluorescence (chinin) at 365 nm Max. 1.0 ppb
Explosion Limit 13.2 %
Vapor Pressure 184 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR