tert-Butylmethylether ≥99,9%, CHROMASOLV™ Plus für die HPLC, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

650560-1L
650560-1LEA 179.52 EUR
650560-1L
tert-Butylmethylether ≥99,9%, CHROMASOLV™ Plus für die HPLC, Riedel-de Haën™
tert-Butylmethylether

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Formel: (CH₃)₃COCH₃
Siedepunkt: 55,3 °C (1.013 hPa)
Schmelzpunkt: –109 °C
Dichte: 0,74 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008812
CAS-Nummer: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity (GC) Min. 99.85 %
IR spectrum complying
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 400 nm Max. 0.005
Absorbance at 300 nm Max. 0.005
Absorbance at 250 nm Max. 0.10
Absorbance at 225 nm Max. 0.50
Absorbance at 210 nm Max. 1.0
Fluorescence (chinin) at 365 nm Max. 1.0 ppb
GC (as heptachlor epoxide) Max. 10 ng/l
Refractive Index n20/D 1.369 (lit.)
Autoignition Temperature 705 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR