(S)-(-)-α-Methoxy-α-(trifluormethyl)phenylessigsäure, Supelco®
Lieferant: Merck
Synonyme:
(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid
M-Clarity™ Qualitätsstufe = MQ100
65369-250MG-F
65369-250MG-FEA
148
EUR
65369-250MG-F
(S)-(-)-α-Methoxy-α-(trifluormethyl)phenylessigsäure, Supelco®
(S)-(-)-α-Methoxy-α-(trifluormethyl)phenylessigsäure
Formel:
C₁₀H₉F₃O₃ Molecular Weight: 234,17 g/mol |
MDL:
MFCD00064200 CAS-Nummer: 17257-71-5 EINECS: 241-292-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...