Tween® 20 (Polysorbat), Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Polyoxyethylene sorbitan monolaurate

655205-250
655205-250EA 105 EUR
655205-250
Tween® 20 (Polysorbat), Millipore®
Tween® 20 (Polysorbat)
Formel: C₅₈H₁₁₄O₂₆
Molecular Weight: 1227,53 g/mol
Dichte: 1,1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00165986
CAS-Nummer: 9005-64-5
EINECS: 500-018-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR