Natriumhydroxid 10.0 M NaOH (10.0N) Volumetric, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Caustic soda
656054-1L
656054-1LEA
47.62
EUR
656054-1L
Natriumhydroxid 10.0 M NaOH (10.0N) Volumetric, Fluka™
Sodium hydroxide
Honeywell's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)Buffers available as concentrates or ready-to-useReagents for complexometry:Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
UNSPSC Code: 12000000
Formel:
NaOH Molecular Weight: 40 g/mol Siedepunkt: 120 °C Schmelzpunkt: 323 Dichte: 1.350 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003548 CAS-Nummer: 1310-73-2 UN: 1824 ADR: 8 II Merck Index: 14,08627 |
Spezifikation Testergebnisse
Assay | 9.80 - 10.20 N |
Carbonate (as Na₂CO₃) | Max. 0.12 N |
Traceable to NIST SRM | effective |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...