N-Methylbistrifluoracetamid, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide, MBTFA
M-Clarity™ Qualitätsstufe = MQ100
65943-25ML
65943-5ML
65943-25MLEA
1210
EUR
65943-25ML
65943-5ML
N-Methylbistrifluoracetamid, Supelco®
N-Methylbistrifluoracetamid
Formel:
C₅H₃F₆NO₂ Molecular Weight: 223.07 g/mol |
MDL:
MFCD00000412 CAS-Nummer: 685-27-8 EINECS: 211-680-5 UN: 2920 ADR: 8.3 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...