N-Methylbistrifluoracetamid, Supelco®

Lieferant: Merck
Gefahr

Synonyme: 2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide, MBTFA

M-Clarity™ Qualitätsstufe = MQ100

65943-25ML 65943-5ML
65943-25MLEA 1210 EUR
65943-25ML 65943-5ML
N-Methylbistrifluoracetamid, Supelco®
N-Methylbistrifluoracetamid
Formel: C₅H₃F₆NO₂
Molecular Weight: 223.07 g/mol
MDL: MFCD00000412
CAS-Nummer: 685-27-8
EINECS: 211-680-5
UN: 2920
ADR: 8.3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82d68c7c0030edd294c4d779d4c6a6ba924161a8&vid=34aeb6cca37b41a107d7b2f46cc78e077c2ff307&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR