Organic reference standard, Hydroxycitronellal, analytical standard, Supelco®
Lieferant: Merck
66010-1ML
66010-1MLEA 46.5 EUR
66010-1ML
Organic reference standard, Hydroxycitronellal, analytical standard, Supelco®
Standards Organische Standards
Hydroxycitronellal is a synthetic fragrance material used in perfumes. It can be prepared from citronellal either by hydration of citronellal bisulfite or by hydrolysis with sulphuric acid and alkali.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f46229c6792459acb07360489283fffbb0245a24&vid=f46229c6792459acb07360489283fffbb0245a24&prid=web&clid=SAPDAM