pH software, EasyDirect™
Lieferant: Mettler - Toledo
30323214
661-0271EA 247 EUR
661-0271
pH software, EasyDirect™
Software Labor-Software
This intuitive software for improved data management can be used with SevenCompact™, SevenCompact™ Duo, Seven2Go™ Pro, FiveEasyPlus or SevenExcellence™ pH meters. Within a few clicks the data measured on the pH meter can be seamlessly transferred to a computer. The application will store the data unmodified and allow for further printing or analysis.
  • Available in 12 languages
  • Data can be stored, printed or exported to Excel or any other csv application
  • Up to three instruments can be connected simultaneously
Bestellinformation: Supplied without meter, computer and USB cable.