Doppelplatinstift-Elektrode, DM143-SC
Lieferant: Mettler - Toledo
51107699
661-1002EA 0 EUR
661-1002
Doppelplatinstift-Elektrode, DM143-SC
Elektroden Elektroden, sonstige
The DM143-SC is a durable, double-pin platinum electrode requiring zero maintenance. Primarily used for redox titrations of food constituents such as Vitamine C or free and total SO2 in wine.