pH/mV/°C/ion meters, bench, 780 / 781
Lieferant: METROHM
2.780.0010 2.781.0010
662-0378EA 10920 EUR
662-0378 662-0410
pH/mV/°C/ion meters, bench, 780 / 781
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
Top-level instruments with the highest degree of comfort.
  • Multi-point calibration with up to max. 9 buffers
  • Memory possibility for 100 measuring values with date, time and sample identification
  • Automatic electrode test (graphical printout) for pH electrodes
  • Automatic temperature compensation and automatic buffer recognition
  • 781: Ion measurement by direct measurement or automatic standard/sample addition or subtraction with Dosimat Plus (optional)
Bestellinformation: 780 and 781 supplied with Unitrode pH electrode, stand rod with base, electrode holder and line adapter.