Buffer solutions
Lieferant: Mettler - Toledo
51302068
51302080
51302047
51302069
51302070
51302079
662-0540EA
64.25
EUR
662-0540
662-0544
662-0538
662-0541
662-0542
662-0543
Buffer solutions
Puffer
pH-Puffer
Solutions in sachets ensure optimum precision.
- Advantages
- No danger of contamination
The plastic-coated sachet provides a perfect liquid seal and good protection from light. The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards. They have been adjusted using a pH-metre that has been calibrated with a buffer associated with NIST standards.
Lieferumfang: The buffers in sachets are delivered with a certificate of conformity.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...