Filling solutions for pH electrodes, Orion™
Lieferant: Thermo Orion
900003
900019
510011
810007
900001
900002
900004
900011
910008-WA
662-1524EA
189
EUR
662-1524
ORIO900019
662-1555
662-0717
662-0749
662-0750
662-0751
662-0753
ORIO910008-WA
Filling solutions for pH electrodes, Orion™
Elektrolyte
Elektrodenfülllösungen
Filling solutions for pH and reference electrodes.
- ROSS filling solution for use only with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
- When the single junction reference electrode or double junction reference electrodes are used with Orion ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide