Electrode extension cables, Orion™
Lieferant: Thermo Orion
910027
662-0769EA 206 EUR
662-0769
Electrode extension cables, Orion™
Kabel
These cables can be used with a variety of electrodes.
  • Extend the length of an electrode cable by 15 ft. (4,57 m)
  • Different connectors available