pH electrode, micro, Orion™ 9826BN
Lieferant: Thermo Orion
9826BN
662-0888EA 845 EUR
662-0888
pH electrode, micro, Orion™ 9826BN
Elektroden pH-Elektroden
This long length combination electrode can be used to accurately measure pH in NMR cells.
  • Made of very thin, delicate glass
  • Refillable