pH/mV/ORP/°C meters, bench top, Orion Star™ A211
Lieferant: Thermo Orion
STARA2110 STARA2115 STARA2116 STARA2114 STARA2117 STARA2119
662-1442EA 880 EUR
662-1442 662-1443 ORIOSTARA2116 ORIOSTARA2114 ORIOSTARA2117 ORIOSTARA2119
pH/mV/ORP/°C meters, bench top, Orion Star™ A211
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
Easy to use and budget friendly instrument for a wide range of applications and advanced pH analysis in the lab. Ideal for simple, routine pH, mV and temperature measurements.
  • Performs up to a 5-point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Eliminate the need for magnetic stirring bars using the meter-controlled stirrer probe to uniformly mix solutions
  • Data log collects and date/time stamps up to 2000 measurement sets with optional sample and user ID
  • Calibration log stores the 10 most recent calibrations
  • Non volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star™ Com software facilitates data transfer to a computer via USB or RS232 connections
  • IP 54 rated housing resists dust and splashes and can be wall-mounted if table space is limited
Capture your critical readings with measurement stability indicator and selectable read modes AUTO-READ™ holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals.
Use the included electrode stand to easily place sensors in samples and simplify storage to minimise breakage.
Standards: CE, TUV 3-in-1 and FCC Class A.
Lieferumfang: All meters are supplied with electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate.
Kit 1 additionally includes 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD stainless steel ATC probe, 810199 ROSS pH buffer and solution kit.
Kit 2 additionally includes 8302BNUMD ROSS Ultra Triode glass-body pH/ATC electrode, 810199 ROSS pH buffer and solution kit.
Kit 3 additionally includes 8157BNUMD ROSS Ultra Triode epoxy-body pH/ATC electrode, 810199 ROSS pH buffer and solution kit.
Kit 4 additionally includes 9157BNMD Triode epoxy-body pH/ATC electrode, 910199 pH buffer and solution kit.
Kit 5 additionally includes 9165BNWP Sure-Flow epoxy-body pH electrode, 927007MD stainless steel ATC probe, 910199 pH buffer and solution kit.