pH/mV/°C meter, handheld, Orion Star™ A321
Lieferant: Thermo Orion
STARA3210 STARA3215
662-1446EA 860 EUR
662-1446 662-1447
pH/mV/°C meter, handheld, Orion Star™ A321
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This portable pH meter is designed a wide range of pH, mV, ORP and temperature testing and field applications. The meter is suitable for use in the most demanding locations thanks to its IP 67-rated housing.
  • Up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
  • Calibration log stores ten most recent calibrations
  • Non volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
Portable meter offers an information-rich screen and user-friendly operation. This rugged, waterproof portable meter is ready to tackle the element and designed to be easy-to-use, dependable field meters that offer advanced features and functions including calibration and high/low limit alarms and password-protected measurement methods.
Standards: TUV 3-in-1 and FCC Class A certifications.
Lieferumfang: Supplied with four AA batteries, literature CD, printed quick start guide, computer interface cable, meter test certificate and 3-year replacement warranty.
The kit additionally includes: 8107UWMMD ROSS Ultra Triode gel filled epoxy-body pH/ATC electrode (3 m cable), Orion pH 4/7/10 buffer pouches (10 pack each), ROSS electrode storage solution (475 ml), Orion rinse solution pouches (10 pack), Orion Star A series portable meter armor, and Orion Star A series hard-sided field case.