ATC probes, glass body, Orion™
Lieferant: Thermo Orion
927006MD
662-1458EA 278 EUR
662-1458
ATC probes, glass body, Orion™
Elektroden pH-Elektroden
Automatic Temperature Compensation (ATC) probes for fast, accurate measurements in the lab or field.
  • 927006MD automatically corrects pH measurements for variation in electrode slope due to temperature change
  • With glass body: Designed for use in organic solvents
  • Compatible with Orion meters
Electrode with MiniDIN connector is designed for use with Orion Star series meters.
Electrode with 3,5 mm phono tip connector is designed for use with the 310, 320, 330, 350 and 370 PerpHecT meters as well as the 162A, 535A, 550A and 555A meters.