Combination pH electrode, Orion™ 9156BNWP
Lieferant: Thermo Orion
9156BNWP
662-1531EA 184 EUR
662-1531
Combination pH electrode, Orion™ 9156BNWP
Elektroden pH-Elektroden
This general purpose electrode is ideal for performing routine pH measurements under rugged conditions.
  • Unbreakable body extends beyond the pH sensing glass bulb for protection
  • With waterproof connector
Lieferumfang: Supplied with 60 ml bottle of Orion electrode fill solution.