Combination pH electrode, Orion™ Sure-Flow™ 9165BNWP
Lieferant: Thermo Orion
9165BNWP
662-1532EA 273 EUR
662-1532
Combination pH electrode, Orion™ Sure-Flow™ 9165BNWP
Elektroden pH-Elektroden
Combination pH electrode with epoxy body for fast response in difficult samples and soils.
  • Refillable Ag/AgCl combination pH electrode
  • Waterproof BNC connector
Compatible with: Thermo Scientific™ Orion Star™ Series meters and competitive meter models with BNC connection.