Storage solution for ROSS electrodes
Lieferant: Thermo Orion
810001
662-1548EA 145 EUR
662-1548
Storage solution for ROSS electrodes
Elektrolyte Elektrodenaufbewahrungslösungen
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH electrode storage solutions.
  • Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples