Orion™ 9502BNWP Carbon Dioxide Electrode
Lieferant: Thermo Orion
9502BNWP
662-1591EA 890 EUR
662-1591
Orion™ 9502BNWP Carbon Dioxide Electrode
Elektroden ISE-Elektroden
This electrode measures carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly and accurately.
  • Gas-sensing combination electrode
  • With waterproof BNC connector