VWR® pHenomenal® IS 2100 L, pH-/mV-/°C-/Ionen-Messgerät, Tischmodell

Lieferant: VWR Collection
662-1658EA 2010 EUR
662-1658
VWR® pHenomenal® IS 2100 L, pH-/mV-/°C-/Ionen-Messgerät, Tischmodell
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
Dieses benutzerfreundliche Instrument mit Elektroden-Gelenkstativ und Gehäuse der Schutzart IP 43 bietet eine hohe Auflösung und Genauigkeit für präzise Multiparameter-Messungen. Mit USB-Ausgang zum Anschluss an einen Computer. GLP-konform.

  • Large graphic LCD with continuous LED backlight shows both pH/mV and temperature value simultaneously
  • Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets
  • Automatic power shut-off: Adjustable from 10 min to 24 h
  • Automatic buffer recognition of DIN and NIST buffers (1,68/4,00/6,86/9,18/12,54)
  • Three additional technical buffers at 25 °C (4,00/7,00/10,00) in memory
  • Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
  • Low energy consumption. Choice of mains supply or 4× AA batteries for over 1000 operating hours
  • Low battery indicator

Lieferumfang: Ready to use IS 2100L kit includes instrument, glass pH electrode with built-in temperature sensor 221, 1 m cable and articulated stand, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, short instructions and CD ROM with manual.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR