pH-Kombinationselektroden
Lieferant: Mettler - Toledo
51109503 51109505 51109507 51109502 51109500 51109504 51109500 51109502
662-2161EA 615 EUR
662-2161 662-2163 662-1894 662-2160 662-2159 662-1669 662-2475 662-2478
pH-Kombinationselektroden
Elektroden pH-Elektroden
Combined glass pH electrodes for direct pH measurement and acid/base titrations in aqueous or non-aqueous media. The all-rounder for a large variety of uncritical applications in almost all kinds of industries: Acid content of beverages, acid and base capacity of water samples, pH start applications.
  • Integrated glass sleeve for secure fixation in the titration head
  • With S7 screw cap and detachable cap or with Plug & Play S12 connection
  • Plug & Play sensor features an integrated sensor chip which allows data storing and monitoring
DG111-SC/DGi111-SC/DG114-SC/DGi114-SC: ARGENTHAL™ Referenzsystem
DG113-SC/DGi113-SC: Ag/AgCl/LiCl Referenzsystem
DGi101-SC/DG115-SC/DGi115-SC: ARGENTHAL™ Referenzsystem mit Silberionenfalle
DGi116-Solvent: ARGENTHAL™/LiCl Referenzsystem
Bestellinformation: Each electrode is supplied with 25 ml of reference electrolyte, individual quality certificate and unique serial number.