Pocket ORP tester, Thermo Scientific™ Eutech™ Elite ORP
Lieferant: EUTECH
ELITEORP
662-2302EA 215 EUR
662-2302
Pocket ORP tester, Thermo Scientific™ Eutech™ Elite ORP
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This rugged, user-friendly tester features a simultaneous display of the redox measurement with temperature. Oxidation-Reduction Potential (ORP) or redox potential measurements are used to monitor chemical reactions, to quantify ion activity, or to determine the oxidising or reducing properties of a solution.
  • Intuitive keypad and large graphic LCD with backlight
  • Auto-hold readings; one-push calibration
  • Easy to replace sensor
  • Leakproof cap with fill-line indicator
  • Wide, stable base allows the tester to stand independently
  • Lanyard provision for ‘on the go’ measurements
  • IP 67 waterproof rating
Typical applications include: Hydroponics, food and beverages, environmental, drinking water, wastewater, agriculture, aquaculture and aquariums, pools and spas, education and industrial (cooling towers).
Warranty: 1 year
Lieferumfang: Supplied with sensor, lanyard, batteries and manual.