pH/ion meter, SevenDirect SD50
Lieferant: Mettler - Toledo
30671544 30671545 30671546 30671547 30671548 30671549 30671552
662-2384EA 1720 EUR
662-2384 662-2385 662-2386 662-2387 662-2388 662-2389 662-2392
pH/ion meter, SevenDirect SD50
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
Robust bench top meter for accurate pH and ion determinations in the lab and near production.
  • Large colour touch screen provides direct measurement and easy data entry
  • Instructions guide users through the sensor calibration process
  • Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)
The large colour touch screen offers direct menu access, fast data entry and guides users through routine tasks. The instruments supports GLP compliance by storing time stamp, sample, sensor and method IDs with every measurement